Mouse JAM-C/JAM3 PicoKine; ELISA Kit, Boster
Catalog # 76466-662
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Pack type:Plastic box
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Size:96 wells/kit, with removable strips
- Storage Conditions:Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
- Storage Temperature:Store at 4 °C for 6 months, at −20 °C for 12 months.
- Cat. No.:76466-662
Specifications
About this item
Sandwich high sensitivity ELISA kit for quantitative detection of mouse JAM-C/JAM3. 96 wells/kit, with removable strips.