Order Entry
United States
ContactUsLinkComponent
Formalin concentrate 4X (10% final concentration) neutral buffered, Epredia™
Corrosive
Toxic
Health
Formalin concentrate 4X (10% final concentration) neutral buffered, Epredia™
Catalog # 84000-020
Supplier:  Epredia
CAS Number:  
undefined
Corrosive
Toxic
Health
Formalin concentrate 4X (10% final concentration) neutral buffered, Epredia™
Catalog # 84000-020
Supplier:  Epredia
Supplier Number:  5700
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle
  • Size:
    1 gal.
  • Cat. No.:
    84000-020
  • Supplier no.:
    5700

Specifications

About this item

 :
ADR:9,
 :30.03 g/mol
UN:3334
 :GHS05
 :GHS06
 :GHS08
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274