Order Entry
United States
ContactUsLinkComponent
Polyethylene glycol 200 viscous liquid
Polyethylene glycol 200 viscous liquid
Catalog # 100504-204
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Polyethylene glycol 200 viscous liquid
Catalog # 100504-204
Supplier Number:  19700
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    450 ml
  • Cat. No.:
    100504-204
  • Supplier no.:
    19700

Specifications

About this item

 :
 :200 g/mol
Density:1.2390 (20 °C)
CAS Number:25322-68-3
 :MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779596