ContactUsLinkComponent
HOBOnet Temperature and Humidity Sensor
HOBOnet Temperature and Humidity Sensor
Catalog # 470323-658
undefined
HOBOnet Temperature and Humidity Sensor
Catalog # 470323-658
Supplier Number:  RXW-THC-900
Specifications
  • Description:
    HOBOnet Temp/RH sensor
  • Cat. No.:
    470323-658
  • Accuracy:
    T: ±0.2 °C, RH: ±2.5%
  • Range:
    T: –40...75 °C; RH: 0 - 100%
Specifications
About this item
The HOBOnet Wireless Temperature and Relative Humidity Sensor provides a cost-effective and scalable solution for web-enabled monitoring of air temperature and humidity.
  • 450 to 600 meter wireless range and up to 5 extended hops with a wireless repeater
  • Simple push button setup for joining a HOBOnet wireless network
  • Powered by rechargeable AA batteries and built–In solar panel or lithium batteries
  • Operates in an outdoor environment with a solar radiation shield
Ordering information: A complete system requires at least one HOBO RX3000 Remote Monitoring Station, a HOBOnet Wireless Manager, and a HOBOnet Wireless Sensor, OR one HOBO MicroRX Station (which has an integrated HOBOnet Wireless Manager) and a HOBOnet Wireless Sensor. HOBOnet Wireless Repeaters can be added to extend the range.