Venturi General Purpose Bench Fume Hoods, Kewaunee
Supplier: Avantor
Certificates
About this item
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features including: Adjustable LED lights, electromechanical sash stop, ultra-smooth sash system and push-button sash management.
- Energy efficient
- Low flow/high performance
- Large viewing height
- LED interior lighting
- Venturi post design improves corner airflow
Color: Light neutral or snow white
Working height: Sash 28"
Frequently Bought Together