ContactUsLinkComponent
MicroMax® Disposable Protective​ Coveralls with Elastic Wrists and Attached Hood and Boots, Lakeland
MicroMax® Disposable Protective​ Coveralls with Elastic Wrists and Attached Hood and Boots, Lakeland
Catalog # 75856-884
Supplier:  Lakeland
undefined
MicroMax® Disposable Protective​ Coveralls with Elastic Wrists and Attached Hood and Boots, Lakeland
Catalog # 75856-884
Supplier:  Lakeland
Supplier Number:  TG414-XL
Specifications
  • Clothing Size:
    XL
  • Description:
    MicroMax®​ coverall
  • Cat. No.:
    75856-884
  • Supplier no.:
    TG414-XL
Specifications
About this item
Durable coveralls provide protection from dirt, grease, grime, and light chemical splash. Garment also acts as a blood and viral barrier.
  • SMS back panel increases breathablity and provides a barrier to particulates and aerosol mist
  • Elastic back waist provides improved comfort
  • Front and sides material provides protection from dirt, liquids, and light chemical splash
  • Storm flap over zipper protects against splashes
MicroMax® brand disposable protective garments bring the most advanced microporous technology to the protective clothing market. This line of general-purpose protective clothing can be used in work environments where hazardous or non-hazardous contaminants may be present, as well as non-hazardous environments where dirt, grime, splashes, and spills are occurring.
MicroMax® brand fabrics are comprised of a microporous film with a polyester ripstop scrim between the film and substrate that gives this material additional strength for use on the toughest jobs.
Certifications: ASTM F1670. ASTM F1671.