Order Entry
United States
ContactUsLinkComponent
Pliers, Terminal Plug Forming, Excelta
Pliers, Terminal Plug Forming, Excelta
Catalog # 63043-020
Supplier:  Excelta
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Pliers, Terminal Plug Forming, Excelta
Catalog # 63043-020
Supplier:  Excelta
Supplier Number:  907-101
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Pliers, misc. former, 5 star quality, for terminal plugs
  • Length:
    150 mm (6")
  • Material:
    Carbon
  • Cat. No.:
    63043-020
  • Supplier no.:
    907-101

Specifications

About this item

Designed to insert or extract the hard to reach terminal plugs.

  • Forms a terminal plug
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use

The 907-101 five star 6" terminal handling plier is designed to insert or extract the hard to reach terminal plugs. The inside jaw has a cutout to prevent nicking or pulling on the lead wire. It features soft cushioned ergonomic grips (10¹⁰ ohms/sq surface resistivity) with dual leaf springs for smooth action.