ContactUsLinkComponent
Innovating Science® Engineer and Explore Materials for Prothetics
Innovating Science® Engineer and Explore Materials for Prothetics
Catalog # 470329-182
Supplier:  Ward's Science
undefined
Innovating Science® Engineer and Explore Materials for Prothetics
Catalog # 470329-182
Supplier:  Ward's Science
Supplier Number:  IS3404
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Engineer and explore materials for prostetics
  • Cat. No.:
    470329-182
  • No. of uses:
    15
Specifications
About this item
Learn about the materials that biomedical engineers use when creating prosthetic devices.
  • Students will investigate the physical properties of various metals, ceramics, and polymers and then determine which materials would be most suitable for use in a hip replacement prosthetic. Students will also engineer their own prosthetic arm.
  • Aligned to NGSS Practices
  • Performance expectations: MS-ETS1-2, MS-ETS1-3, HS-ETS1-2, HS-ETS1-3
  • Disciplinary core ideas: ETS1.B, ETS1.C
  • Cross cutting concepts include influence of science enginerring and technology on society and the natural world
  • Engineering practices include engaging in argument from evidence, analyzing and interpreting data, constructing explanations and designing solutions
In this lab, students will explore the materials that biomedical engineers use when creating prosthetic devices. They will first investigate the physical properties of various metals, ceramics, and polymers to understand how they differ, and determine which materials would be most suitable for use in a hip replacement prosthetic. Students will then engineer their own prosthetic arm that meets certain design requirements and mimics the functionality of a real hand. Kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copy masters are included.
Delivery: Kit includes instructional manual and SDS booklet.
Caution: WARNING: children under 8 yrs. can choke or suffocate on uninflated or broken balloons. Adult supervision required. Keep uninflated balloons from children. Discard broken balloons at once.