Alethia® Pertussis DNA amplification assay, Meridian Bioscience
Catalog # 76437-460
Supplier: Meridian
CAS Number:
Specifications
- Description:Alethia® Pertussis
- Cat. No.:76437-460
- Details:Molecular assay for the detection of Bordetella pertussis
Specifications
About this item
The Alethia™ Pertussis DNA amplification assay is a qualitative in vitro diagnostic test for the direct detection of Bordetella pertussis in human nasopharyngeal swab samples taken from patients suspected of having respiratory tract infection attributable to Bordetella pertussis.
- With molecular results in less than one hour, long turnaround times and send out costs can be eliminated
- Targeted detection can help physicians treat patients appropriately
The molecular accuracy of Alethia™ Pertussis provides a clear and objective evaluation of positive/negative test results. This can help to reduce the risk of spreading illness to others and allow for appropriate treatment as soon as possible.