Order Entry
United States
ContactUsLinkComponent
VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/conductivity/oxygen), Bench
VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/conductivity/oxygen), Bench
Supplier:  VWR International
Certificates
undefined
VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/conductivity/oxygen), Bench
Supplier:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 76460-494
  • Model:
  • MU 6100L S1
  • pH Range:
  • −2.000...+19.999
  • pH Resolution:
  • 0.1/0.01/0.001 (selectable)
  • pH Accuracy:
  • ±0.1/±0.01/±0.005
  • pH Calibration:
  • Up to 5 points
  • Automatic Buffer Recognition:
  • BDH Prolabo® TITRINORM® buffers at 25 °C: 4.00/6.98/9.94
    NIST/DIN 19266 (1.68/4.00/6.86/9.18/12.54)
    Technical buffers at 25 °C: 4.00/7.00/10.00
  • mV range:
  • ±1200.0/±2500
  • mV resolution:
  • 0.1/1
  • mV accuracy:
  • ±0.3/±1
  • Conductivity Range:
  • 0.001 µS/cm - 1000 mS/cm
  • Conductivity Resolution:
  • 0.1 µS/cm
  • Conductivity Accuracy:
  • ±0.5% of measured value
  • Conductivity Cell Constant:
  • 0.001 - 25 cm⁻¹ (depending on probe)
  • Resistivity accuracy:
  • 1 Ω/cm - 999 MΩ/cm (±0.5% of measured value)
  • Salinity Range:
  • 0.00 - 70 ppt
  • TDS Range:
  • 0 - 1999 mg/L / 2.00 - 19.99 g/L / 20.0 - 199.9 g/L
  • TDS Resolution:
  • 1 mg/L / 0.01 g/L / 0.1 g/L
  • DO Concentration:
  • 0.00 - 20.00 / 0.0 - 90.0 mg/L
  • DO Resolution:
  • 0.1%; 0.01 mg/L (depending on sensor)
  • DO Saturation:
  • 0.0 - 200.0%; 0 - 600% (depending on sensor)
  • DO Accuracy:
  • ±0.5% of value (between 5...30 °C)
  • DO Calibration:
  • Saturated air
  • Temperature Range:
  • Auto: −5.0...+10.0 °C; manual: −25...+130 °C
  • Temperature Resolution:
  • 0.1/1 °C
  • Temperature Accuracy:
  • ±0.2
  • Temperature Compensation:
  • Automatic/manual
  • Display:
  • Graphic LCD with LED backlight
  • Power supply:
  • Mains supply or 4× AA batteries
  • Compliance:
  • IP 43/GLP
  • Data Logging - Log Function:
  • Selectable between 1 s and 1 h (selectable intervals: 1 s, 5 s, 10 s, 30 s, 1 min, 5 min, 10 min, 15 min, 30 min, 60 min)
  • Data Logging Capacity:
  • 5000 data sets
  • Connections:
  • BNC for pH/banana for temperature/8-pin for conductivity and DO
  • Outputs:
  • Waterproof USB connection (slave, bidirectional)

Specifications

Related Information

Frequently Bought Together