Human Recombinant CD40 Ligand (from HEK293 Cells)
Catalog # 103790-254
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:1 mg
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human CD40 Ligand, His Tag (Cat. No. CDL-H5248) at 5 μg/mL (100 μL/well) can bind Human CD40, Fc Tag (Cat. No. CD0-H5253) with a linear range of 0.156-5 μg/mL (QC tested).
- Gene ID:P29965-1
- Protein Synonyms:TRAP|CD40L|IMD3|gp39|CD154|CD40LG|IGM|T-BAM|TNFSF5|HIGM1
- Protein/Peptide Name:CD40 Ligand
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:19.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-254
Specifications
About this item
Human CD40 Ligand / TNFSF5 Protein, His Tag, ACROBiosystems