Human Recombinant CD40 Ligand (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human CD40 Ligand / TNFSF5 Protein, Fc Tag (active trimer) (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag (active trimer) (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:FACS analysis shows that Human CD40 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5269) can bind to 293T-CD40 cells surface CD40. The concentration of Human CD40 Ligand is 0.3 μg/mL (Routinely tested). Human CD40 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5269) stimulates secretion of IL-6 by human B cells. The ED50 for this effect is 9.68-11.48 ng/mL in the presence of 5 ng/mL of Recombinant Human IL‑4 (Routinely tested).
- Gene ID:P29965-1
- Protein Synonyms:TRAP|CD40L|IMD3|gp39|CD154|CD40LG|IGM|T-BAM|TNFSF5|HIGM1
- Protein/Peptide Name:CD40 Ligand
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:74.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together