Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD30 (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Human Recombinant CD30 (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant CD30 (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –20 ℃, aviod light
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Anti-CD30 Mab (Human IgG1) at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human CD30, His Tag (Cat. No. CD0-HF2H3) with a linear range of 0.039-0.625 μg/mL (QC tested). 2e5 of Anti-CD30 CAR-293 cells were stained with 100 μL of 0.3 μg/mL FITC-Labeled Human CD30, His Tag (Cat. No. CD0-HF2H3) and negative control protein respectively, FITC signal was used to evaluate the binding activity (QC tested).
  • Protein Synonyms:
    Ki-1|CD30|D1S166E|TNFRSF8
  • Protein/Peptide Name:
    CD30
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    39.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together