Human Recombinant CD19 (from HEK293 cells), PE (Phycoerythrin)
Catalog # 103871-442
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:25 tests
- Tag sequence:His Tag
- Storage Conditions:–20 ℃, aviod light
- Biological Activity:1e6 of the anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human CD19 (20-291), His Tag (Cat. No. CD9-HP2H3). PE Streptavidin was used as negative control (QC tested).
- Gene ID:P15391-1
- Protein Synonyms:CVID3|MGC12802|B4|CD19
- Protein/Peptide Name:CD19
- Molecular Weight:33.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, 0.5% BSA, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103871-442
Specifications
About this item
PE-Labeled Human CD19 (20-291) Protein, His Tag, ACROBiosystems