Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD133 (from HEK293 cells)
Human Recombinant CD133 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human Recombinant CD133 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –70 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human CD133, His Tag (Cat. No. CD3-H52H4) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human CD133 (133-3) Antibody, Human IgG1 with a linear range of 1-16 ng/mL (QC tested).
  • Gene ID:
    O43490-1
  • Protein Synonyms:
    Prominin-1|CD133|PROM1|AC133|PROML1
  • Protein/Peptide Name:
    CD133
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    97.2 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Delivered as bulk protein in a 0.2 μm filtered solution of 50 mM HEPES, 150 mM NaCl, DDM, CHS, pH7.5 with glycerol as protectant.
  • Shipping Temperature:
    Dry Ice

Specifications

Frequently Bought Together