Human Recombinant BLAME (from HEK293 cells), Biotin
Biotinylated Human BLAME / SLAMF8 Protein, His,Avitag™, ACROBiosystems
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Precision Avi
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:Q9P0V8-1
- Protein Synonyms:CD353|SLAMF8|BLAME
- Protein/Peptide Name:BLAME
- Purity:>85% as determined by SDS-PAGE.
- Molecular Weight:27.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Sodium Citrate, 150 mM NaCl, pH5.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together