Human Recombinant BAFFR (from HEK293 cells), Biotin
Supplier: ACRO Biosystems
Certificates
About this item
Biotinylated Human BAFFR / TNFRSF13C Protein, His,Avitag™ (MALS verified), ACROBiosystems
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Precision Avi
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:Q96RJ3-1
- Protein Synonyms:CVID4|BAFFR|BAFF-R|BROMIX|CD268|TNFRSF13C|prolixin
- Protein/Peptide Name:BAFFR
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:10.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together