Human Recombinant B7-H7 (from HEK293 cells)
Catalog # 103790-108
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:100 µg
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human CD28H, Fc Tag (Cat. No. CDH-H5251) at 5 μg/mL (100 μL/well) can bind Human B7-H7, His Tag (Cat. No. B77-H52H5) with a linear range of 0.02-0.313 μg/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Human B7-H7, His Tag (Cat. No. B77-H52H5) at 5 μg/mL (100 μL/well) can bind Human CD28H, Fc Tag (Cat. No. CDH-H5251) with a linear range of 1-20 ng/mL (Routinely tested).
- Gene ID:Q9UM44-1
- Protein Synonyms:B7-H7|B7 Homolog 7|HHLA2
- Protein/Peptide Name:B7-H7
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:38.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-108
Specifications
About this item
Human B7-H7 / HHLA2 Protein, His Tag, ACROBiosystems