Human Recombinant B7-H4 (from HEK293 cells)
Catalog # 103815-464
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:100 µg
- Tag sequence:Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.01 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human B7-H4, Fc Tag, low endotoxin (Cat. No. B74-H5256) at 0.5 μg/mL (100 μL/well) can bind Anti-B7-H4 MAb (Human IgG1) with a linear range of 0.2-8 ng/mL (QC tested).
- Gene ID:Q7Z7D3-1
- Protein Synonyms:B7h.5|B7-H4|B7S1|VTCN1
- Protein/Peptide Name:B7-H4
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:51.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103815-464
Specifications
About this item
Human B7-H4 Protein, Fc Tag, low endotoxin, ACROBiosystems