Human Recombinant B7-H3 (4Ig) (from HEK293 cells), Biotin
Biotinylated Human B7-H3 (4Ig) / B7-H3b Protein, Fc,Avitag™, ACROBiosystems
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:Fc, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human B7-H3 (4Ig), Fc,Avitag (Cat. No. B7B-H82F5) with a linear range of 0.3-2 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human B7-H3 (4Ig), Fc,Avitag (Cat. No. B7B-H82F5) at 1 μg/mL (100 μL/well) on Streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (Routinely tested).
- Gene ID:Q5ZPR3-1
- Protein Synonyms:B7 homolog 3|B7-H3|UNQ309/PRO352|4Ig-B7-H3|PSEC0249|CD276
- Protein/Peptide Name:B7-H3 (4Ig)
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:74.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together