Human Recombinant APRIL (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human APRIL / TNFSF13 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human BCMA, Fc Tag (Cat. No. BC7-H5254) at 5 μg/mL (100 μL/well) can bind Human APRIL, His Tag (Cat. No. APL-H5244) with a linear range of 8-125 ng/mL (QC tested).
- Gene ID:O75888-1
- Protein Synonyms:APRIL|TNFSF13|TALL2|TRDL-1|ZTNF2|TALL-2|CD256
- Protein/Peptide Name:APRIL
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:22.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, Arginine, pH8.0. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together