Human Recombinant Apolipoprotein E (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human Apolipoprotein E / APOE Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Loaded Biotinylated Human TREM2, His,Avitag (Cat. No. TR2-H82E7) on SA Biosensor, can bind Human Apolipoprotein E, His Tag (Cat. No. APE-H5246) with an affinity constant of 37.1 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P02649-1
- Protein Synonyms:Apolipoprotein E|Apo-E|APOE
- Protein/Peptide Name:Apolipoprotein E
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:36.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, 5 mM CHAPS, 2 mM DTT, pH 7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together