Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant ACE2 (from HEK293 cells), Biotin
 
Human Recombinant ACE2 (from HEK293 cells), Biotin

 

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His, Avitag™
  • Storage Conditions:
    –70 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Biotinylated Human ACE2, His,Avitag (Cat. No. AC2-H82E6) captured on Biotin CAP - Series S sensor Chip can bind SARS-CoV-2 S protein (R683A, R685A), His Tag (Cat. No. SPN-C52H4) with an affinity constant of 54.5 nM as determined in a SPR assay (Biacore 8K) (Routinely tested). Loaded Biotinylated Human ACE2, His,Avitag (Cat. No. AC2-H82E6) on SA Biosensor, can bind SARS-CoV-2 S protein (R683A, R685A), His Tag (Cat. No. SPN-C52H4) with an affinity constant of 42.8 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q9BYF1-1
  • Protein Synonyms:
    ACEH|ACE-2|ACE2
  • Protein/Peptide Name:
    ACE2
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    87.2 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Delivered as bulk protein in a 0.2 μm filtered solution of 50 mM Tris, 150 mM NaCl, pH7.5 with glycerol as protectant.
  • Shipping Temperature:
    Dry Ice

 

Frequently Bought Together