Human Recombinant 4-1BB Ligand (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human 4-1BB Ligand / TNFSF9 (71-254) Protein, His,Flag Tag (active trimer) (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Flag Tag (active trimer) (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (Cat. No. 41L-H52D4) at 2 μg/mL (100 μL/well) can bind Human 4-1BB, Fc Tag (Cat. No. 41B-H5258) with a linear range of 0.2-3 ng/mL (QC tested). Loaded Mouse 4-1BB, Fc Tag (Cat. No. 41B-M5258) on Protein A Biosensor, can bind Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (Cat. No. 41L-H52D4) with an affinity constant of 18.6 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Human 4-1BB Ligand (71-254), His,Flag Tag (active trimer) (MALS verified) (Cat. No. 41L-H52D4) induce IL-8 secretion in HT1080 human CD137 cell line. The EC50 for this effect is 0.20-0.44 μg/mL (Routinely tested).
- Gene ID:P41273-1
- Protein Synonyms:CD137L|TNFSF9|4-1BB Ligand
- Protein/Peptide Name:4-1BB Ligand
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:62.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together