Rabbit Recombinant 4-1BB (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Rabbit 4-1BB / TNFRSF9 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rabbit
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human 4‑1BB Ligand, Fc Tag (Cat. No. 41L-H5257) at 2 μg/mL (100 μL/well) can bind Rabbit 4-1BB, His Tag (Cat. No. 41B-R52H9) with a linear range of 1-39 ng/mL (QC tested). Loaded Human 4-1BB Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. 41L-H5254) on Protein A Biosensor, can bind Rabbit 4-1BB, His Tag (Cat. No. 41B-R52H9) with an affinity constant of 2.1 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:G1SHD7-1
- Protein Synonyms:CD137|TNFRSF9|ILA|CDw137|4-1BB
- Protein/Peptide Name:4-1BB
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:18.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together