Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells)
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    SARS-CoV-2
  • Tag sequence:
    Mouse IgG2a Fc Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Loaded SARS-CoV-2 S1 protein, Mouse IgG2a Fc Tag (Cat. No. S1N-C5257) on AMC Biosensor, can bind Human ACE2, His Tag (Cat. No. AC2-H52H8) with an affinity constant of 3.95 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). FACS analysis shows that SARS-CoV-2 S1 protein, Mouse IgG2a Fc Tag (Cat. No. S1N-C5257) can bind to Vero E6 cells surface ACE2. The concentration of SARS-CoV-2 S1 protein is 3 μg/ml. (Routinely tested).
  • Protein Synonyms:
    Spike|Spike glycoprotein Subunit1|S glycoprotein Subunit1|S1 protein|Spike protein S1|COVID-19
  • Protein/Peptide Name:
    S1 protein
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    101.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together