SARS-CoV-2 Recombinant S1 protein (from HEK293 cells), Biotin
Biotinylated SARS-CoV-2 (COVID-19) S1 protein, His,Avitag™ (MALS verified), ACROBiosystems
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Tag sequence:His, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Loaded Biotinylated SARS-CoV-2 S1 protein, His,Avitag (Cat. No. S1N-C82E8) on SA Biosensor, can bind Human ACE2, His Tag (Cat. No. AC2-H52H8) with an affinity constant of 1.61 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Protein Synonyms:Spike|Spike glycoprotein Subunit1|S glycoprotein Subunit1|S1 protein|Spike protein S1|COVID-19
- Protein/Peptide Name:S1 protein
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:78.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together