Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells), Biotin
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells), Biotin
Supplier:  ACRO Biosystems
Certificates
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells), Biotin
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    SARS-CoV-2
  • Tag sequence:
    Precision Avi
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Protein Synonyms:
    Spike|Spike glycoprotein Subunit1|S glycoprotein Subunit1|S1 protein|Spike protein S1|COVID-19
  • Protein/Peptide Name:
    S1 protein
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    78.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together