Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 Recombinant S protein RBD (from HEK293 cells)
SARS-CoV-2 Recombinant S protein RBD (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
SARS-CoV-2 Recombinant S protein RBD (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Storage Conditions:
    –70 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Protein Synonyms:
    Spike|S glycoprotein RBD|Spike glycoprotein Receptor-binding domain|Spike protein RBD|S protein RBD|COVID-19
  • Protein/Peptide Name:
    S protein RBD
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    52.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Delivered as bulk protein in a 0.2 μm filtered solution of 10 mM PB, Arginine, pH7.4.
  • Shipping Temperature:
    Dry Ice

Specifications

Frequently Bought Together