SARS-CoV-2 Recombinant S protein RBD (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
SARS-CoV-2 (COVID-19) S protein RBD, Fc Tag (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Tag sequence:Fc Tag (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Loaded SARS-CoV-2 S protein RBD, Fc Tag (Cat. No. SPD-C5255) on Protein A Biosensor, can bind Human ACE2, His Tag (Cat. No. AC2-H52H8) with an affinity constant of 2.16 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded SARS-CoV-2 S protein RBD, Fc Tag (Cat. No. SPD-C5255) on Protein A Biosensor, can bind Cynomolgus ACE2, His Tag (Cat. No. AC2-C52H7) with an affinity constant of 4.48 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Protein Synonyms:Spike|S glycoprotein RBD|Spike glycoprotein Receptor-binding domain|Spike protein RBD|S protein RBD|COVID-19
- Protein/Peptide Name:S protein RBD
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:51.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together