SARS-CoV-2 Recombinant S protein RBD (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
SARS-CoV-2 (COVID-19) S protein RBD, Mouse IgG2a Fc Tag(MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Tag sequence:Mouse IgG2a Fc Tag (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Loaded SARS-CoV-2 S protein RBD, Mouse IgG2a Fc Tag (Cat. No. SPD-C5259) on AMC Biosensor, can bind Human ACE2, His Tag (Cat. No. AC2-H52H8) with an affinity constant of 3.44 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). FACS analysis shows that the binding of SARS-CoV-2 S protein RBD, Mouse IgG2a Fc Tag (Cat. No. SPD-C5259) to Vero E6 cells surface ACE2 was inhibited by increasing concentration of Anti-SARS-CoV-2 Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35). The concentration of SARS-CoV-2 S protein RBD used is 5μg/ml. The IC50 is 10.33 μg/ml (Routinely tested).
- Protein Synonyms:Spike|S glycoprotein RBD|Spike glycoprotein Receptor-binding domain|Spike protein RBD|S protein RBD|COVID-19
- Protein/Peptide Name:S protein RBD
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:52.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS,pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together