SARS-CoV-2 Recombinant S protein (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
SARS-CoV-2 S protein, His Tag, Super stable trimer (MALS & NS-EM verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Protein Synonyms:Spike|Spike glycoprotein|S protein|S glycoprotein|COVID-19
- Protein/Peptide Name:S protein
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:138.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together