SARS-CoV-2 Recombinant Papain-like Protease (from E. coli)
SARS-CoV-2 (COVID-19) Papain-like Protease Protein, His Tag (active enzyme), ACROBiosystems
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Storage Conditions:–70 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Protein Synonyms:PL-PRO|pp1a|Plpro|Replicase polyprotein 1a|Papain-like Protease|ORF1a polyprotein|1a|COVID-19
- Protein/Peptide Name:Papain-like Protease
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:37.6 kDa
- Endotoxin Level:Low
- Formulation:Delivered as bulk protein in a 0.2 μm filtered solution of 50 mM HEPES, 300 mM NaCl, 1 mM TCEP, pH7.5 with glycerol as protectant.
- Shipping Temperature:Dry Ice
Frequently Bought Together