Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 Recombinant NSP16 & NSP10 (from E. coli)
SARS-CoV-2 Recombinant NSP16 & NSP10 (from E. coli)
Supplier:  ACRO Biosystems
Certificates
SARS-CoV-2 Recombinant NSP16 & NSP10 (from E. coli)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    SARS-CoV-2
  • Storage Conditions:
    –70 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Protein Synonyms:
    NSP16 & NSP10|nsp16 & nsp10|COVID-19
  • Protein/Peptide Name:
    NSP16 & NSP10
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    35.3 kDa (NSP16) & 18.3 kDa (NSP10)
  • Endotoxin Level:
    Low
  • Formulation:
    Delivered as bulk protein in a 0.2 μm filtered solution of PBS, pH7.4 with glycerol as protectant.
  • Shipping Temperature:
    Dry Ice

Specifications

Frequently Bought Together