Order Entry
United States
ContactUsLinkComponent
Rhesus macaque Recombinant VEGF R1 (from HEK293 cells)
Rhesus macaque Recombinant VEGF R1 (from HEK293 cells)
Catalog # 103790-878
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Rhesus macaque Recombinant VEGF R1 (from HEK293 cells)
Catalog # 103790-878
Supplier:  ACROBIOSYSTEMS
Supplier Number:  VE1R52571MG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rhesus monkey
  • Size:
    1 mg
  • Tag sequence:
    Mouse IgG2a Fc Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 0.1 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Rhesus macaque PLGF (Cat. No. PGF-R52H0) at 0.5 μg/mL (100 μL/well) can bind Rhesus macaque VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-R5257) with a linear range of 0.6-10 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Rhesus macaque VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-R5257) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF165, His Tag (Cat. No. VE5-H82Q0) with a linear range of 0.5-3.9 ng/mL (Routinely tested).
  • Gene ID:
    F7FAY2-1
  • Protein Synonyms:
    FLT|FLT1|VEGFR1
  • Protein/Peptide Name:
    VEGF R1
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    109.1 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT
  • Cat. No.:
    103790-878

Specifications

About this item

Rhesus macaque VEGF R1 / Flt-1 Protein, Mouse IgG2a Fc Tag, low endotoxin, ACROBiosystems