Order Entry
United States
ContactUsLinkComponent
 
Rhesus macaque Recombinant Fc gamma RIII/ CD16 (from HEK293 cells)
 
Rhesus macaque Recombinant Fc gamma RIII/ CD16 (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rhesus monkey
  • Tag sequence:
    His Tag (SPR & BLI verified)
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Rhesus macaque CD16, His Tag (Cat. No. FC6-R52H6) captured on CM5 Chip via anti-His antibody can bind Rituximab with an affinity constant of 0.232 μM as determined in SPR assay (Biacore T200) (QC tested). Loaded Rhesus macaque CD16, His Tag (Cat. No. FC6-R52H6) on HIS1K Biosensor, can bind Rituximab with an affinity constant of 0.28 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    A3RFZ7-1
  • Protein Synonyms:
    FCGR3
  • Protein/Peptide Name:
    Fc gamma RIII/ CD16
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    23.1 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

 

Frequently Bought Together