Rabbit Recombinant FcRn (FCGRT & B2M) (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Rabbit FcRn / FCGRT&B2M Heterodimer Protein, His Tag&Tag Free (SPR verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rabbit
- Tag sequence:His Tag&Tag Free (SPR verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Rabbit FCGRT&B2M Heterodimer Protein, His Tag&Tag Free (Cat. No. FCM-R52W5) captured on CM5 Chip via anti-His antibody can bind Herceptin® with an affinity constant of 2.12 μM as determined in SPR assay (Biacore T200) (QC tested).
- Protein Synonyms:FcRn|FCGRT & B2M
- Protein/Peptide Name:FcRn (FCGRT & B2M)
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:32.1 kDa (FCGRT) & 11.7 kDa (B2M)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together