Recombinant Streptavidin (from E. coli), HRP (Horseradish Peroxidase)
Catalog # 103871-566
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:HRP (Horseradish Peroxidase)
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Size:1 mg
- Storage Conditions:–20 °C
- Biological Activity:Immobilized Nivolumab at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Avitag,His Tag (Cat. No. PD1-H82E4) with a linear range of 0.8-13 ng/mL when detected by Streptavidin-HRP (Cat. No. STN-NH115) (QC tested).
- Gene ID:P22629
- Protein Synonyms:Streptavidin|SA
- Protein/Peptide Name:Streptavidin
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Na3PO4 and NaCl pH6.8. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103871-566
Specifications
About this item
Streptavidin-HRP, ACROBiosystems