Order Entry
United States
ContactUsLinkComponent
 
Mouse Recombinant TSLP (from HEK293 cells)
 
Mouse Recombinant TSLP (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    Q9JIE6-1
  • Protein/Peptide Name:
    TSLP
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    15.9 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

 

Frequently Bought Together