Mouse Recombinant Properdin (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Mouse Properdin Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P11680-1
- Protein Synonyms:CFP|PFC|Complement factor P|Properdin
- Protein/Peptide Name:Properdin
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:49.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together