Mouse Recombinant LAIR-1 (from HEK293 Cells)
Catalog # 103790-636
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Mouse
- Size:1 mg
- Tag sequence:Mouse IgG2a Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Mouse LAIR-1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LA1-M5254) with a linear range of 1-10 ng/mL (QC tested).
- Gene ID:Q8BG84-1
- Protein Synonyms:LAIR1|CD305
- Protein/Peptide Name:LAIR-1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:40.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-636
Specifications
About this item
Mouse LAIR1 / CD305 Protein, Mouse IgG2a Fc Tag, low endotoxin, ACROBiosystems