Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant Integrin alpha V beta 8 (from HEK293 Cells)
Mouse Recombinant Integrin alpha V beta 8 (from HEK293 Cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Mouse Recombinant Integrin alpha V beta 8 (from HEK293 Cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    His Tag&Tag Free
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Mouse ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-M52W6) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His,Avitag (Cat. No. TG1-H82Qb) with a linear range of 1-10 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Human Fibronectin at 2 μg/mL (100 μL/well) can bind Mouse ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-M52W6) with a linear range of 0.156-5 μg/mL (Routinely tested). Measured by its binding ability in a functional ELISA. Immobilized Human Vitronectin at 2 μg/mL (100 μL/well) can bind Mouse ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT8-M52W6) with a linear range of 0.156-5 μg/mL (Routinely tested).
  • Gene ID:
    P43406-1 (ITGAV) & Q0VBD0-1 (ITGB8)
  • Protein Synonyms:
    ITGAV&ITGB8|Integrin alpha V beta 8
  • Protein/Peptide Name:
    Integrin alpha V beta 8
  • Purity:
    >95% as determined by reduced SDS-PAGE.
  • Molecular Weight:
    112.5 kDa (ITGAV) & 77.5 kDa (ITGB8)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together