Mouse Recombinant IL-3 R alpha (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Mouse IL-3 R alpha / CD123 Protein, His Tag (SPR verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag (SPR verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Mouse IL-3 R alpha, His Tag (Cat. No. ILA-M52H4) captured on CM5 Chip via anti-His antibody can bind Mouse IL-3, Tag Free with an affinity constant of 0.704 μM as determined in SPR assay (Biacore T200) (QC tested). Loaded Mouse IL-3 R alpha, His Tag (Cat. No. ILA-M52H4) on NTA Biosensor, can bind Mouse IL-3, Tag Free with an affinity constant of 0.38 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P26952-1
- Protein Synonyms:IL3RX|IL3RA|CD123 antigen|IL-3Ra|IL3RY|CD123|IL-3 R alpha|hIL3Ra|MGC34174|hIL-3Ra|IL3RAY|IL3R|IL-3R-alpha
- Protein/Peptide Name:IL-3 R alpha
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:36.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together