Mouse Recombinant Glypican 1 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Mouse Glypican 1 / GPC1 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 1 μg/mL (100 μL/well) can bind Mouse Glypican 1, His Tag (Cat. No. GP1-M52H7) with a linear range of 5-78 ng/mL (QC tested).
- Gene ID:Q9QZF2-1
- Protein Synonyms:Glypican 1|GPC1
- Protein/Peptide Name:Glypican 1
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:57.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together