Mouse Recombinant Fc gamma RIII/ CD16 (from HEK293 Cells)
Catalog # 103815-520
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Mouse
- Size:50 µg
- Tag sequence:His Tag (SPR & BLI verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Mouse CD16, His Tag (SPR & BLI verified) (Cat. No. CDA-M52H8) Captured on NTA-Series S sensor chip can bind OKT3 with an affinity constant of 0.126 μM as determined in a SPR assay (Biacore T200) (QC tested). Loaded Mouse CD16, His Tag (SPR & BLI verified) (Cat. No. CDA-M52H8) on NTA Biosensor, can bind OKT3 with an affinity constant of 0.78 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Mouse CD16, His Tag (SPR & BLI verified) (Cat. No. CDA-M52H8) on NTA Biosensor, can bind Herceptin with an affinity constant of 2.8 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q5D5I8
- Protein Synonyms:FCGR3
- Protein/Peptide Name:Fc gamma RIII/ CD16
- Purity:>95% as determined by reduced SDS-PAGE.
- Molecular Weight:23.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103815-520
Specifications
About this item
Mouse Fc gamma RIII / CD16 Protein, His Tag (SPR & BLI verified), ACROBiosystems