Influenza Recombinant Hemagglutinin (HA) (from HEK293 cells)
Supplier: ACRO Biosystems
Certificates
About this item
Influenza A[A/swine/Jiangsu/J004/2018(H1N1)] Hemagglutinin (HA) Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Influenza
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Protein Synonyms:HA|Hemagglutinin
- Protein/Peptide Name:Hemagglutinin (HA)
- Molecular Weight:63.3 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT
Specifications
Frequently Bought Together