Human Recombinant CD24 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Human CD24 Protein, Mouse IgG2a Fc Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Mouse IgG2a Fc Tag
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human CD24, Mouse IgG2a Fc Tag (Cat. No. CD4-H5257) at 5 μg/mL (100 μL/well) can bind Anti-CD24 MAb (SN3) with a linear range of 20-313 ng/mL (QC tested).
- Gene ID:P25063-1
- Protein Synonyms:CD24A|CD24
- Protein/Peptide Name:CD24
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:30.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together