Human Recombinant Vitronectin (from E. coli), UltraLys
Supplier: ACRO Biosystems
Certificates
About this item
Biotinylated Human Vitronectin / VTN Protein, Tag Free, ultra sensitivity (primary amine labeling), ACROBiosystems
Specifications
- Conjugation:UltraLys
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:Tag Free
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Biotinylated Human Vitronectin, Tag Free, primary amine labeling (Cat. No. VIN-H8113) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human ITGAV&ITGB5 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT5-H52W5) with a linear range of 0.156-1.25 μg/mL (QC tested).
- Gene ID:P04004-1
- Protein Synonyms:Serum-spreading factor|Vitronectin|VTN|S-protein|V75|VN
- Protein/Peptide Name:Vitronectin
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:43.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together