Human Recombinant VEGF110 (from HEK293 Cells)
Catalog # 103790-874
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:500 µg
- Tag sequence:Tag Free (HPLC-verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized ActiveMax® Human VEGF110, Tag Free (HPLC-verified) (Cat. No. VE0-H5212) at 0.05 μg/mL (100 μL/well) can bind Human VEGF R1, His Tag (Cat. No. VE1-H5220) with a linear range of 4-63 ng/mL (QC tested). ActiveMax® Human VEGF110, Tag Free (HPLC-verified) (Cat. No. VE0-H5212) stimulates proliferation of human umbilical vein endothelial cells (HUVEC). The EC50 for this effect is 5.06-11.59 ng/mL (Routinely tested).
- Gene ID:P15692-4
- Protein Synonyms:RP1-261G23.1|MVCD1|MGC70609|VEGFA|VPF
- Protein/Peptide Name:VEGF110
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:12.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103790-874
Specifications
About this item
ActiveMax® Human VEGF110 Protein, Tag Free (HPLC-verified), ACROBiosystems