Human Recombinant TIGIT (from HEK293 Cells)
103815-518
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:100 µg
- Tag sequence:His Tag (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Anti-Human TIGIT MAb (Mouse IgG1) captured on CM5 chip via Anti-Mouse antibodies surface, can bind Human TIGIT, His Tag (Cat. No. TIT-H52H3) with an affinity constant of 3.93 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded Anti-Human TIGIT MAb (Mouse IgG1) on AMC Biosensor, can bind Human TIGIT, His Tag (Cat. No. TIT-H52H3) with an affinity constant of 3.72 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human CD155, Fc Tag (Cat. No. CD5-H5251) on Protein A Biosensor, can bind Human TIGIT, His Tag (Cat. No. TIT-H52H3) with an affinity constant of 0.23 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). Loaded Human Nectin-3, Fc Tag (Cat. No. PV3-H5255) on Protein A Biosensor, can bind Human TIGIT, His Tag (Cat. No. TIT-H52H3) with an affinity constant of 0.732 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). FACS assay shows that Human TIGIT, His Tag (Cat. No. TIT-H52H3) can bind to 293T cell overexpressing human CD155. The concentration of TIGIT used is 0.3 μg/mL (Routinely tested).
- Gene ID:Q495A1-1
- Protein Synonyms:VSTM3|TIGIT|VSIG9
- Protein/Peptide Name:TIGIT
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:14.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103815-518
Human TIGIT Protein, His Tag (MALS verified), ACROBiosystems